Learn More
Biomatik Corporation Pig Ciliary Neurotrophic Factor (CNTF) ELISA Kit
Supplier:  Biomatik Corporation EKU03193
Encompass_Preferred
Pig Ciliary Neurotrophic Factor (CNTF) ELISA Kit for detecting CNTF in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O02732. Target Synonyms: HCNTF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4800