Learn More
Biomatik Corporation Pig Clusterin (CLU) ELISA Kit
Supplier:  Biomatik Corporation EKU10727
Encompass_Preferred
Pig Clusterin (CLU) ELISA Kit for detecting CLU in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.69ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q29549. Gene ID: 397025. Target Synonyms: CLU; TRPM2; KUB1; CLI; APOJ; SGP2; AAG4; APO-J; SP-40; Apolipoprotein J; Complement Lysis Inhibitor; 40,Sulfated Glycoprotein 2; Testosterone-Repressed Prostate Message 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6897