Learn More
American Research Products Inc Pig coagulation factor VIII F
Catalog No. 5000329345
Encompass
Pig coagulation factor VIII (FVIII) ELISA Kit from CUSABIO Detection Range 62 5 ng/mL-4000 ng/mL Sensitivity 15 62 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29345 Supplier American Research Products Inc Supplier No. CSBE15000P
Pig coagulation factor VIII (FVIII) ELISA Kit from CUSABIO Detection Range 62 5 ng/mL-4000 ng/mL Sensitivity 15 62 ng/mL Sample serum plasma tissue homogenates Method Competitive