Learn More
Biomatik Corporation Pig Collagen Type IV (COL4) ELISA Kit
Supplier:  Biomatik Corporation EKU10735
Encompass_Preferred
Pig Collagen Type IV (COL4) ELISA Kit for detecting COL4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 6.25-400ng/ml. Sensitivity: 2.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: Type-IV Collagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4602