Learn More
American Research Products Inc Pig complement fragment 5a C5
Catalog No. 5000327361
Encompass
Pig complement fragment 5a (C5a) ELISA kit from CUSABIO Detection Range 9 375 ng/mL-600 ng/mL Sensitivity 2 34 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27361 Supplier American Research Products Inc Supplier No. CSBEQ027637PI
Pig complement fragment 5a (C5a) ELISA kit from CUSABIO Detection Range 9 375 ng/mL-600 ng/mL Sensitivity 2 34 ng/mL Sample serum plasma tissue homogenates Method Sandwich