Learn More
Biomatik Corporation Pig Connective Tissue Growth Factor (CTGF) CLIA Kit
Supplier:  Biomatik Corporation EKU09526
Encompass_Preferred
Pig Connective Tissue Growth Factor (CTGF) CLIA Kit for detecting CTGF in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.27-200ng/ml. Sensitivity: 0.13ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O19113. Target Synonyms: CCN2; HCS24; IGFBP8; NOV2; Insulin-Like Growth Factor-Binding Protein 8; Hypertrophic Chondrocyte-Specific Protein 24; CCN Family Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8504