Learn More
Biomatik Corporation Pig Copeptin (CPP) ELISA Kit
Supplier:  Biomatik Corporation EKU12129
Encompass_Preferred
Pig Copeptin (CPP) ELISA Kit for detecting CPP in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.45pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01183. Gene ID: 396995. Target Synonyms: C-Terminal Provasopressin; Vasopressin-Neurophysin 2-Copeptin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5356