Learn More
Biomatik Corporation Pig C-Peptide (CP) ELISA Kit
Supplier:  Biomatik Corporation EKU08348
Encompass_Preferred
Pig C-Peptide (CP) ELISA Kit for detecting CP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 6.17-500pg/ml. Sensitivity: 2.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01315. Gene ID: 397415. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6414