Learn More
American Research Products Inc Pig creatine kinase MB isoenzy
Catalog No. 5000327509
Encompass
Pig creatine kinase MB isoenzyme (CK-MB) ELISA kit from CUSABIO Detection Range 2 35 mU/mL-150 mU/mL Sensitivity 0 58 mU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27509 Supplier American Research Products Inc Supplier No. CSBEQ027653PI
Pig creatine kinase MB isoenzyme (CK-MB) ELISA kit from CUSABIO Detection Range 2 35 mU/mL-150 mU/mL Sensitivity 0 58 mU/mL Sample serum plasma tissue homogenates Method Sandwich