Learn More
Biomatik Corporation Pig Creatine Kinase, Muscle (CKM) ELISA Kit
Supplier:  Biomatik Corporation EKU10752
Encompass_Preferred
Pig Creatine Kinase, Muscle (CKM) ELISA Kit for detecting CKM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5XLD3. Gene ID: 397264. Target Synonyms: CK-MM; CKMM; CK3; CK-M; MM-CK; M-CK; Creatine kinase M-type, N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8297