Learn More
American Research Products Inc Pig Endothelial nitric oXide s
Catalog No. 5000329733
Encompass
Pig Endothelial nitric oXide synthase eNOS ELISA Kit from CUSABIO Detection Range 3 12 IU/mL-200 IU/mL Sensitivity 0 78 IU/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29733 Supplier American Research Products Inc Supplier No. CSBE08574P
Pig Endothelial nitric oXide synthase eNOS ELISA Kit from CUSABIO Detection Range 3 12 IU/mL-200 IU/mL Sensitivity 0 78 IU/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich