Learn More
American Research Products Inc Pig Ferritin heaVy chain FTH1
Catalog No. 5000327654
Encompass
Pig Ferritin heaVy chain (FTH1) ELISA kit from CUSABIO Detection Range 4 7 ng/ml- 300 ng/ml Sensitivity 1 175 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27654 Supplier American Research Products Inc Supplier No. CSBEL009030PI
Pig Ferritin heaVy chain (FTH1) ELISA kit from CUSABIO Detection Range 4 7 ng/ml- 300 ng/ml Sensitivity 1 175 ng/ml Sample serum plasma tissue homogenates Method Sandwich