Learn More
American Research Products Inc Pig Fibrinogen Degradation Pro
Catalog No. 5000328756
Encompass
Pig Fibrinogen Degradation Product FDP ELISA Kit from CUSABIO Detection Range 1 25 g/mL-80 g/mL Sensitivity 0 31 g/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28756 Supplier American Research Products Inc Supplier No. CSBE06531P
Pig Fibrinogen Degradation Product FDP ELISA Kit from CUSABIO Detection Range 1 25 g/mL-80 g/mL Sensitivity 0 31 g/mL Sample serum plasma tissue homogenates Method Sandwich