Learn More
American Research Products Inc Pig fibroblast growth factor 1
Catalog No. 5000327036
Encompass
Pig fibroblast growth factor 19 (FGF19) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-27036 Supplier American Research Products Inc Supplier No. CSBE17583P
Pig fibroblast growth factor 19 (FGF19) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich