Learn More
Biomatik Corporation Pig Fibroblast Growth Factor 21 (FGF21) CLIA Kit
Supplier:  Biomatik Corporation EKU09537
Encompass_Preferred
Pig Fibroblast Growth Factor 21 (FGF21) CLIA Kit for detecting FGF21 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.98pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: C6JWG4. Gene ID: 100302504. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8501