Learn More
American Research Products Inc Pig Follicle stimulating hormo
Catalog No. 5000326556
Encompass
Pig Follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 20 mIU/mL-700 mIU/mL Sensitivity 10 mIU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26556 Supplier American Research Products Inc Supplier No. CSBE06791P
Pig Follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 20 mIU/mL-700 mIU/mL Sensitivity 10 mIU/mL Sample serum plasma tissue homogenates Method Sandwich