Learn More
American Research Products Inc Pig glial fibrillary acidic pr
Catalog No. 5000329715
Encompass
Pig glial fibrillary acidic protein GFAP ELISA Kit from CUSABIO Detection Range 0 1 ng/mL-20 ng/mL Sensitivity 0 05 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29715 Supplier American Research Products Inc Supplier No. CSBE14950P
Pig glial fibrillary acidic protein GFAP ELISA Kit from CUSABIO Detection Range 0 1 ng/mL-20 ng/mL Sensitivity 0 05 ng/mL Sample serum plasma tissue homogenates Method Competitive