Learn More
Biomatik Corporation Pig Glutathione Peroxidase 3, Plasma (GPX3) ELISA Kit
Supplier:  Biomatik Corporation EKU10871
Encompass_Preferred
Pig Glutathione Peroxidase 3, Plasma (GPX3) ELISA Kit for detecting GPX3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 7.8-500ng/ml. Sensitivity: 3.2ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A287AIJ3. Target Synonyms: GSHPX3; GPx-P; GSHPx-P; GPXP; Plasma glutathione peroxidase; Extracellular glutathione peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7633