Learn More
American Research Products Inc Pig growth hormone GH ELISA K
Catalog No. 5000329629
Encompass
Pig growth hormone GH ELISA Kit from CUSABIO Detection Range 2 5 ng/mL-100 ng/mL Sensitivity 2 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29629 Supplier American Research Products Inc Supplier No. CSBE06813P
Pig growth hormone GH ELISA Kit from CUSABIO Detection Range 2 5 ng/mL-100 ng/mL Sensitivity 2 5 ng/mL Sample serum plasma tissue homogenates Method Competitive