Learn More
Biomatik Corporation Pig Growth Hormone Releasing Hormone (GHRH) ELISA Kit
Supplier:  Biomatik Corporation EKU04619
Encompass_Preferred
Pig Growth Hormone Releasing Hormone (GHRH) ELISA Kit for detecting GHRH in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 12.35-1000pg/ml. Sensitivity: 5.22pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: E0Y439. Gene ID: 100499556. Target Synonyms: GRF; GHRF; Somatocrinin; Growth-Hormone-Releasing Factor; Sermorelin; Somatoliberin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5858