Learn More
Immunology Consultants Pig Haptoglobin ELISA Kit
SDP
Supplier:  Immunology Consultants E5HPT
Encompass_Preferred
The Pig HPT test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring HPT in pig biological samples.
Catalog No. 50-240-6554