Learn More
American Research Products Inc Pig Heat Shock Protein 70 HSP
Catalog No. 5000330208
Encompass
Pig Heat Shock Protein 70 HSP-70 ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30208 Supplier American Research Products Inc Supplier No. CSBE08317P
Pig Heat Shock Protein 70 HSP-70 ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich