Learn More
Biomatik Corporation Pig Heat Shock Protein 90kDa Beta 1 (HSP90b1) ELISA Kit
Supplier:  Biomatik Corporation EKU12402
Encompass_Preferred
Pig Heat Shock Protein 90kDa Beta 1 (HSP90b1) ELISA Kit for detecting HSP90b1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q29092. Gene ID: 397191. Target Synonyms: HSP90-B1; ECGP; GP96; GRP94; TRA1; Glycoprotein 96; Tumor Rejection Antigen (gp96) 1;94 kDa glucose-regulated protein; Endoplasmin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5258