Learn More
American Research Products Inc Pig hepatocyte growth factor
Catalog No. 5000329137
Encompass
Pig hepatocyte growth factor HGF ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29137 Supplier American Research Products Inc Supplier No. CSBE06795P
Pig hepatocyte growth factor HGF ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich