Learn More
MSC PIG High-Visibility Roll, 24 in. x 75 ft., 1 ea.
Catalog No. 5000765951
Encompass
The PIG® Chat Mat® High-Visibility Absorbent Roll is designed to clean up floor spills while displaying a safety message. This high-visibility yellow mat roll features a repeating safety sign image, making it suitable for aisles and walkways.
  • Wide mediumweight rolls absorb everyday spills, leaks, and overspray.
  • Bright yellow color enhances visibility, alerting workers to hazards.
  • Durable top layer resists scuffs and withstands foot traffic.
  • Safety message repeats throughout the roll to promote a safer work area.
  • Mat lasts 2x longer than competitive mats, resisting ripping or tearing.
  • Easy-tear perforations allow for reducing waste.
  • Absorbs common industrial liquids: oils, coolants, solvents, and water.
  • Can be wrung out and incinerated for waste reduction or fuels blending.
Catalog No. 50-007-65951 Supplier MSC Supplier No. 91458539
The PIG® Chat Mat® High-Visibility Absorbent Roll is designed to clean up floor spills while displaying a safety message. This high-visibility yellow mat roll features a repeating safety sign image, making it suitable for aisles and walkways.
  • Wide mediumweight rolls absorb everyday spills, leaks, and overspray.
  • Bright yellow color enhances visibility, alerting workers to hazards.
  • Durable top layer resists scuffs and withstands foot traffic.
  • Safety message repeats throughout the roll to promote a safer work area.
  • Mat lasts 2x longer than competitive mats, resisting ripping or tearing.
  • Easy-tear perforations allow for reducing waste.
  • Absorbs common industrial liquids: oils, coolants, solvents, and water.
  • Can be wrung out and incinerated for waste reduction or fuels blending.