Learn More
Biomatik Corporation Pig Hypoxia Inducible Factor 1 Alpha (HIF1a) CLIA Kit
Supplier:  Biomatik Corporation EKU09505
Encompass_Preferred
Pig Hypoxia Inducible Factor 1 Alpha (HIF1a) CLIA Kit for detecting HIF1a in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.54pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: A0T1H6. Gene ID: 396696. Target Synonyms: HIF1-A; MOP1; PASD8; Basic Helix-Loop-Helix Transcription Factor; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; Class E basic helix-loop-helix protein 78. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8508