Learn More
Innovative Research Inc Pig Hypxa Indcibl FI Alpha Kit
Supplier:  Innovative Research Inc IPCHIF1AKT
Encompass_Preferred
Porcine Hypoxia Inducible Factor 1 Alpha ELISA Kit - This kit is for the quantitative determination of Porcine Hypoxia Inducible Factor 1 Alpha in biofluid samples, such as serum, plasma, tissue homogenates and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . HIF1a present in samples reacts with an anti-Porcine HIF1a capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-HIF1a detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active HIF1a in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Porcine HIF1a. For research use only.
Catalog No. 50-274-4122