Learn More
Bethyl Laboratories, Inc Pig Igg Elisa Kit
Supplier:  Bethyl Laboratories, Inc E101104
Encompass_Preferred
Pig IgG ELISA Kit, 1 x pre-coated 96-well strip plate, standard, detection antibody, sample dilution buffer, HRP solution, TMB substrate, stop solution, 20X wash buffer, sealing tape
Catalog No. NC0364994