Learn More
Biomatik Corporation Pig Immunoglobulin E (IgE) CLIA Kit
Supplier:  Biomatik Corporation EKU11643
Encompass_Preferred
Pig Immunoglobulin E (IgE) CLIA Kit for detecting IgE in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: K7ZJQ0. Target Synonyms: IGHE; Immunoglobulin Heavy Constant Epsilon; Ig epsilon chain C region. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8109