Learn More
Biomatik Corporation Pig Immunoglobulin G (IgG) CLIA Kit
Supplier:  Biomatik Corporation EKU08634
Encompass_Preferred
Pig Immunoglobulin G (IgG) CLIA Kit for detecting IgG in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.17-300ug/ml. Sensitivity: 0.51ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6182