Learn More
American Research Products Inc Pig Immunoglobulin G IgG ELI
Catalog No. 5000326451
Encompass
Pig Immunoglobulin G (IgG) ELISA Kit from CUSABIO Detection Range 0 586 g/mL-150 g/mL Sensitivity 0 467 g/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-26451 Supplier American Research Products Inc Supplier No. CSBE06804P
Pig Immunoglobulin G (IgG) ELISA Kit from CUSABIO Detection Range 0 586 g/mL-150 g/mL Sensitivity 0 467 g/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive