Learn More
Biomatik Corporation Pig Insulin Like Growth Factor Binding Protein 2 (IGFBP2) ELISA Kit
Supplier:  Biomatik Corporation EKU04991
Encompass_Preferred
Pig Insulin Like Growth Factor Binding Protein 2 (IGFBP2) ELISA Kit for detecting IGFBP2 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.469-30ng/ml. Sensitivity: 0.18ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24853. Gene ID: 397064. Target Synonyms: IBP2; IGF-BP53. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4472