Learn More
American Research Products Inc Pig Interferon b IFN b IFNB E
Catalog No. 5000326229
Encompass
Pig Interferon b IFN-b/IFNB ELISA Kit from CUSABIO Detection Range 50 pg/mL-2000 pg/mL Sensitivity 25 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26229 Supplier American Research Products Inc Supplier No. CSBE09890P
Pig Interferon b IFN-b/IFNB ELISA Kit from CUSABIO Detection Range 50 pg/mL-2000 pg/mL Sensitivity 25 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich