Learn More
American Research Products Inc Pig Interleukin 1 receptor ant
Catalog No. 5000329128
Encompass
Pig Interleukin 1 receptor antagonist IL-1ra ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29128 Supplier American Research Products Inc Supplier No. CSBE04946P
Pig Interleukin 1 receptor antagonist IL-1ra ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich