Learn More
American Research Products Inc Pig Interleukin 12 IL 12 P40
Catalog No. 5000329085
Encompass
Pig Interleukin 12 IL-12/P40 ELISA KIT from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-29085 Supplier American Research Products Inc Supplier No. CSBE11341P
Pig Interleukin 12 IL-12/P40 ELISA KIT from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates Method Sandwich