Learn More
American Research Products Inc Pig Interleukin 12 IL 12 P70
Catalog No. 5000329010
Encompass
Pig Interleukin 12 IL-12/P70 ELISA Kit from CUSABIO Detection Range 0 45 ng/mL-30 ng/mL Sensitivity 0 11 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29010 Supplier American Research Products Inc Supplier No. CSBE13288P
Pig Interleukin 12 IL-12/P70 ELISA Kit from CUSABIO Detection Range 0 45 ng/mL-30 ng/mL Sensitivity 0 11 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich