Learn More
American Research Products Inc Pig Interleukin 22 IL 22 ELI
Catalog No. 5000330151
Encompass
Pig Interleukin 22 (IL-22) ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30151 Supplier American Research Products Inc Supplier No. CSBE15847P
Pig Interleukin 22 (IL-22) ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich