Learn More
American Research Products Inc Pig Interleukin 23 subunit alp
Catalog No. 5000326889
Encompass
Pig Interleukin-23 subunit alpha (IL23A) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26889 Supplier American Research Products Inc Supplier No. CSBEL011638PI
Pig Interleukin-23 subunit alpha (IL23A) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich