Learn More
Biomatik Corporation Pig Interleukin 8 (IL8) CLIA Kit
Supplier:  Biomatik Corporation EKU08809
Encompass_Preferred
Pig Interleukin 8 (IL8) CLIA Kit for detecting IL8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 27.4-20,000pg/ml. Sensitivity: 12.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P26894. Gene ID: 396880. Target Synonyms: CXCL8; AMCF-I; GCP1; K60; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP1; SCYB8; TSG1; B-ENAP; Neutrophil-Activating Protein 1; Granulocyte Chemotactic Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8625