Learn More
American Research Products Inc Pig Keratinocyte Growth Factor
Catalog No. 5000326673
Encompass
Pig Keratinocyte Growth Factor KGF ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200pg/mL Sensitivity 0 78 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26673 Supplier American Research Products Inc Supplier No. CSBE06800P
Pig Keratinocyte Growth Factor KGF ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200pg/mL Sensitivity 0 78 pg/mL Sample serum plasma Method Sandwich