Learn More
American Research Products Inc Pig luteinizing hormone LH E
Catalog No. 5000326250
Encompass
Pig luteinizing hormone (LH) ELISA kit from CUSABIO Detection Range 0 156 ng/ml-10 ng/ml Sensitivity 0 039/ml Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26250 Supplier American Research Products Inc Supplier No. CSBE12870P
Pig luteinizing hormone (LH) ELISA kit from CUSABIO Detection Range 0 156 ng/ml-10 ng/ml Sensitivity 0 039/ml Sample serum plasma tissue homogenates Method Competitive