Learn More
American Research Products Inc Pig Lysyl OXidase ELISA Kit
Catalog No. 5000330138
Encompass
Pig Lysyl OXidase ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30138 Supplier American Research Products Inc Supplier No. CSBE15813P
Pig Lysyl OXidase ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich