Learn More
Cusabio Technology LLC Pig Lysyl Oxidase ELISA Kit
Catalog No. 5000052036
Encompass_Preferred
This kit detects LOX (Lysyl Oxidase) in pig-derived samples including serum plasma and tissue homogenates with a detection range of 1 56 ng/mL to 100 ng/mL and a sensitivity of 0 39 ng/mL Utilizing a quantitative sandwich ELISA methodology it provides reliable measurements within 1-5 hours The assay requires a sample volume of 50-100 L and measures absorbance at 450 nm Designed for accurate quantification of LOX it is ideal for studies involving extracellular matrix remodeling fibrosis or connective tissue disorders in porcine models The kit ensures high specificity and reproducibility making it suitable for both research and diagnostic applications requiring precise LOX level assessment in biological matrices
Catalog No. 50-000-52036 Supplier Cusabio Technology LLC Supplier No. CSBE15813P
This kit detects LOX (Lysyl Oxidase) in pig-derived samples including serum plasma and tissue homogenates with a detection range of 1 56 ng/mL to 100 ng/mL and a sensitivity of 0 39 ng/mL Utilizing a quantitative sandwich ELISA methodology it provides reliable measurements within 1-5 hours The assay requires a sample volume of 50-100 L and measures absorbance at 450 nm Designed for accurate quantification of LOX it is ideal for studies involving extracellular matrix remodeling fibrosis or connective tissue disorders in porcine models The kit ensures high specificity and reproducibility making it suitable for both research and diagnostic applications requiring precise LOX level assessment in biological matrices