Learn More
Biomatik Corporation Pig Macrophage Migration Inhibitory Factor (MIF) ELISA Kit
Supplier:  Biomatik Corporation EKU11058
Encompass_Preferred
Pig Macrophage Migration Inhibitory Factor (MIF) ELISA Kit for detecting MIF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80928. Gene ID: 397412. Target Synonyms: GIF; GLIF; MMIF; Glycosylation-Inhibiting Factor; L-dopachrome isomerase; L-dopachrome tautomerase; Phenylpyruvate tautomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8236