Learn More
American Research Products Inc Pig major acute phase protein
Catalog No. 5000329396
Encompass
Pig major acute phase protein (MAP) ELISA Kit from CUSABIO Detection Range 37 5 ng/mL-2400 ng/mL Sensitivity 9 38 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29396 Supplier American Research Products Inc Supplier No. CSBE13425P
Pig major acute phase protein (MAP) ELISA Kit from CUSABIO Detection Range 37 5 ng/mL-2400 ng/mL Sensitivity 9 38 ng/mL Sample serum plasma tissue homogenates Method Competitive