Learn More
Biomatik Corporation Pig Matrix Metalloproteinase 10 (MMP10) CLIA Kit
Supplier:  Biomatik Corporation EKU09565
Encompass_Preferred
Pig Matrix Metalloproteinase 10 (MMP10) CLIA Kit for detecting MMP10 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.55-400pg/ml. Sensitivity: 0.22pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: C9VZX5. Gene ID: 396769. Target Synonyms: STMY2; SL2; Stromelysin 2; Transin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7074