Learn More
Biomatik Corporation Pig Matrix Metalloproteinase 13 (MMP13) CLIA Kit
Supplier:  Biomatik Corporation EKU09576
Encompass_Preferred
Pig Matrix Metalloproteinase 13 (MMP13) CLIA Kit for detecting MMP13 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.07-50ng/ml. Sensitivity: 0.03ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: F1SV56. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7394