Learn More
Biomatik Corporation Pig Myoglobin (MYO) CLIA Kit
Supplier:  Biomatik Corporation EKU08872
Encompass_Preferred
Pig Myoglobin (MYO) CLIA Kit for detecting MYO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.27-200ng/ml. Sensitivity: 0.10ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02189. Gene ID: 397467. Target Synonyms: PVALB; MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5056