Learn More
Biomatik Corporation Pig Natriuretic Peptide Precursor B (NPPB) ELISA Kit
Supplier:  Biomatik Corporation EKU11897
Encompass_Preferred
Pig Natriuretic Peptide Precursor B (NPPB) ELISA Kit for detecting NPPB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07634. Gene ID: 396844. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8067