Learn More
American Research Products Inc Pig neutrophil gelatinase asso
Catalog No. 5000328623
Encompass
Pig neutrophil gelatinase-associated lipocalin (NGAL) ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28623 Supplier American Research Products Inc Supplier No. CSBE16507P
Pig neutrophil gelatinase-associated lipocalin (NGAL) ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Competitive